Please login to your online account to display your discounted pricing

1-[5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Piperazin-1-yl)pyridine-5-boronic acid pinacol ester
CAS 871125-86-9
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C15H24BN3O2
Formula Weight 289.18g/mol
MDL Number MFCD06795677
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383292500

  SDS
thermo fisher scientific
383292500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White to off-white
Melting Point 138° to 143°C
Quantity 250mg
Chemical Name or Material 2-(Piperazin-1-yl)pyridine-5-boronic acid pinacol ester
CAS 871125-86-9
CAS Min % 96.0
CAS Max % 100.0
Physical Form White to off-white powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C15H24BN3O2
Formula Weight 289.18g/mol
MDL Number MFCD06795677
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.