Please login to your online account to display your discounted pricing

1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW06046EA

 View more versions of this product

Catalog No. SEW06046EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 239.2
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C12H8F3NO
CAS 156496-70-7
MDL Number MFCD01567130
Packaging Amber Glass Bottle
Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde

Specifications

Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
CAS 156496-70-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H8F3NO
Formula Weight 239.2
MDL Number MFCD01567130