Please login to your online account to display your discounted pricing

1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde, Maybridge

Chemical Identifiers

Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde
CAS 156496-70-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H8F3NO
Formula Weight 239.2
MDL Number MFCD01567130
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW06046EA

  SDS
fisher scientific company
SEW06046EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 239.2
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C12H8F3NO
CAS 156496-70-7
MDL Number MFCD01567130
Packaging Amber Glass Bottle
Chemical Name or Material 1-[4-(Trifluoromethyl)phenyl]-1H-pyrrole-2-carbaldehyde