Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[4-(Trifluoromethoxy)phenyl]ethanol, 97%

$83.93 - $83.93

Chemical Identifiers

Chemical Name or Material 1-[4-(Trifluoromethoxy)phenyl]ethanol
CAS 1737-28-6
Molecular Formula C9H9F3O2
Formula Weight 206.16
MDL Number MFCD07784263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3156106

  SDS
alfa aesar™
H3156106
5g Each for $83.93
Description & Specifications

Specifications

Formula Weight 206.16
Chemical Name or Material 1-[4-(Trifluoromethoxy)phenyl]ethanol
Quantity 5g
Assay 97%
CAS 1737-28-6
Molecular Formula C9H9F3O2
MDL Number MFCD07784263