Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-tert-Butylphenyl)homopiperazine dihydrochloride, 98%

$430.24 - $430.24

Chemical Identifiers

Chemical Name or Material 1-(4-tert-Butylphenyl)homopiperazine dihydrochloride
CAS 934992-03-7
Molecular Formula C15H24N2·2HCl
Formula Weight 305.30g/mol
MDL Number MFCD08436124
Synonym 4-BPHP·2HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51702ME

 
alfa aesar™
H51702-ME
Each for $430.24
Description & Specifications

Specifications

Melting Point 240° to 244°C
Quantity 500mg
Chemical Name or Material 1-(4-tert-Butylphenyl)homopiperazine dihydrochloride
CAS 934992-03-7
Assay 98%
Molecular Formula C15H24N2·2HCl
Formula Weight 305.30g/mol
MDL Number MFCD08436124
Synonym 4-BPHP·2HCl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.