missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-tert-Butylbenzyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5532006

 View more versions of this product

Catalog No. AAH5532006



Specifications

Specifications

1-(4-tert-Butylbenzyl)piperazine
White to Yellow
5g
C15H24N2
1-4-t-butylbenzyl piperazine
CC(C)(C)C1=CC=C(C=C1)CN2CCNCC2
232.371
232.37
97%
Air Sensitive
52°C to 55°C
956-61-6
MFCD00082594
UQLCETYSARZZSR-UHFFFAOYSA-N
1-[(4-tert-butylphenyl)methyl]piperazine
2735657
Solid

Chemical Identifiers

956-61-6
232.371
UQLCETYSARZZSR-UHFFFAOYSA-N
2735657
CC(C)(C)C1=CC=C(C=C1)CN2CCNCC2
C15H24N2
MFCD00082594
1-4-t-butylbenzyl piperazine
1-[(4-tert-butylphenyl)methyl]piperazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

Documents