Please login to your online account to display your discounted pricing

1-[4-(tert-Butyl)phenyl]ethan-1-one, 97%, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  BTB09870EA

 View more versions of this product

Catalog No. BTB09870EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 176.26
Quantity 10g
Chemical Name or Material 1-[4-(tert-Butyl)phenyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C12H16O
CAS 943-27-1

Specifications

Chemical Name or Material 1-[4-(tert-Butyl)phenyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C12H16O
Formula Weight 176.26