Please login to your online account to display your discounted pricing

1-[4-(tert-Butyl)phenyl]ethan-1-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-[4-(tert-Butyl)phenyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C12H16O
Formula Weight 176.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

BTB09870DA

  SDS
fisher scientific company
BTB09870DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BTB09870EA

  SDS
fisher scientific company
BTB09870EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.26
Quantity 1g
CAS 943-27-1
Chemical Name or Material 1-[4-(tert-Butyl)phenyl]ethan-1-one
Assay Percent Range 97%
Molecular Formula C12H16O