Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-tert-Butoxyphenyl)homopiperazine monohydrochloride, 98%

$430.24 - $430.24

Chemical Identifiers

Chemical Name or Material 1-(4-tert-Butoxyphenyl)homopiperazine monohydrochloride
CAS 934991-96-5
Molecular Formula C15H24N2O·HCl
Formula Weight 284.83g/mol
MDL Number MFCD16251545
Synonym 4-BOPH·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51687ME

 
alfa aesar™
H51687-ME
Each for $430.24
Description & Specifications

Specifications

Melting Point 197° to 201°C
Quantity 500mg
Chemical Name or Material 1-(4-tert-Butoxyphenyl)homopiperazine monohydrochloride
CAS 934991-96-5
Assay 98%
Molecular Formula C15H24N2O·HCl
Formula Weight 284.83g/mol
MDL Number MFCD16251545
Synonym 4-BOPH·HCl