missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole, TCI America™

Manufacturer:  TCI America P22101G

 View more versions of this product

Catalog No. P22101G



Specifications

Specifications

1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole
147°C
499793-84-9
JFTYJFYWQQLLFG-UHFFFAOYSA-N
1-(4-propylphenyl)-2,5-dithiophen-2-ylpyrrole
132274863
≥98.0% (GC)
Yellow
1g
C21H19NS2
CCCC1=CC=C(C=C1)N2C(=CC=C2C3=CC=CS3)C4=CC=CS4
349.51
Crystal-Powder at 20°C

Chemical Identifiers

499793-84-9
349.51
132274863
CCCC1=CC=C(C=C1)N2C(=CC=C2C3=CC=CS3)C4=CC=CS4
C21H19NS2
JFTYJFYWQQLLFG-UHFFFAOYSA-N
1-(4-propylphenyl)-2,5-dithiophen-2-ylpyrrole
Safety and Handling

Safety and Handling

TSCA : No

Documents