Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Piperidinyloxy)acetylpyrrolidine, 98%

$111.95 - $391.76

Chemical Identifiers

Chemical Name or Material 1-(4-Piperidinyloxy)acetylpyrrolidine
CAS 902836-88-8
Molecular Formula C11H20N2O2
Formula Weight 212.29
MDL Number MFCD08061028
Synonym 4-(2-Oxo-2-pyrrolidin-1-ylethoxy)piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50852MD

  SDS
alfa aesar™
H50852MD
250mg Each for $111.95

AAH5085203

  SDS
alfa aesar™
H5085203
1g Each for $391.76
Description & Specifications

Specifications

Formula Weight 212.29
Synonym 4-(2-Oxo-2-pyrrolidin-1-ylethoxy)piperidine
CAS 902836-88-8
Molecular Formula C11H20N2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-(4-Piperidinyloxy)acetylpyrrolidine
Assay 98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD08061028
Quantity 250mg
Melting Point 63° to 65°C

Applications
It is employed as intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.