missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Phenylenediamine-2-sulfonic Acid 98.0+%, TCI America™

Supplier:  TCI America P169125G

 View more versions of this product

Catalog No. P169125G



Description

Description

Specifications

Specifications

1,4-Phenylenediamine-2-sulfonic Acid
300°C
MFCD00007904
benzenesulfonic acid, 2,5-diamino, p-phenylenediaminesulfonic acid, 2,5-diaminobenzenesulphonic acid, 2,5-diaminobenzene sulfonic acid, 2-sulfo-p-phenylenediamine, ccris 2884, 1,4-phenylenediamine-2-sulfonic acid, unii-up8112d78n, 2,5-diamino benzene sulfonic acid, pubchem13625
[K+].NC1=CC=C(N)C(=C1)S([O-])(=O)=O
226.29
188.20
Crystalline Powder
88-45-9
C6H7KN2O3S
25 g
WRFPRPDYXWUOEK-UHFFFAOYSA-M
potassium 2,5-diaminobenzene-1-sulfonate
66617
≥98.0% (HPLC,T)

Chemical Identifiers

88-45-9
226.29
WRFPRPDYXWUOEK-UHFFFAOYSA-M
66617
[K+].NC1=CC=C(N)C(=C1)S([O-])(=O)=O
C6H7KN2O3S
MFCD00007904
benzenesulfonic acid, 2,5-diamino, p-phenylenediaminesulfonic acid, 2,5-diaminobenzenesulphonic acid, 2,5-diaminobenzene sulfonic acid, 2-sulfo-p-phenylenediamine, ccris 2884, 1,4-phenylenediamine-2-sulfonic acid, unii-up8112d78n, 2,5-diamino benzene sulfonic acid, pubchem13625
potassium 2,5-diaminobenzene-1-sulfonate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2058

RTECSNumber : DB5900100

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions