Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Phenylenediacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 1,4-Phenylenediacetonitrile
CAS 622-75-3
Molecular Formula C10H8N2
Formula Weight 156.19
MDL Number MFCD00001923
UN Number UN3439
Beilstein 1866157
Synonym 1,4-Bis(cyanomethyl)benzene; p-Xylylene dicyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1346414

 
alfa aesar™
L1346414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1346406

 
alfa aesar™
L1346406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.19
Chemical Name or Material 1,4-Phenylenediacetonitrile
Molecular Formula C10H8N2
MDL Number MFCD00001923
TSCA TSCA
Assay 97%
CAS 622-75-3
UN Number UN3439
Beilstein 1866157
Synonym 1,4-Bis(cyanomethyl)benzene; p-Xylylene dicyanide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Melting Point 94° to 98°C
Health Hazard 3 GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Quantity 25g

Applications
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Safety and Handling

Danger

GHS P Statement
P280h-P309-P310
Wear protective gloves/clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.