Please login to your online account to display your discounted pricing

1,4-Pentadien-3-ol, 98%, stabilized, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC426240250

 View more versions of this product

Catalog No. AC426240250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 84.12
Quantity 25g
Color Clear colorless
Density 0.8650g/mL
CAS Max % 100.0
Linear Formula (CH2CH)2CHOH
CAS 922-65-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Molecular Formula C5H8O
Chemical Name or Material 1, 4-Pentadien-3-ol
CAS Min % 97.5
Assay Percent Range 98%
MDL Number MFCD00008633
Name Note Stabilized
Stabilizer approximately 0.4% hydroquinone
Packaging Glass bottle
Synonym Divinyl carbinol
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1, 4-Pentadien-3-ol
Name Note Stabilized
CAS 922-65-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C5H8O
Linear Formula (CH2CH)2CHOH
Formula Weight 84.12
MDL Number MFCD00008633
Synonym Divinyl carbinol
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.