Learn More
1,4-Pentadien-3-ol, 98%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426240050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 4-Pentadien-3-ol | |
922-65-6 | |
100.0 | |
115.0°C | |
Authentic | |
Glass bottle | |
1.4440 to 1.4460 (20°C, 589nm) | |
MFCD00008633 | |
0.865 | |
ICMWSAALRSINTC-UHFFFAOYSA-N | |
penta-1,4-dien-3-ol | |
approximately 0.4% hydroquinone | |
84.12 |
Stabilized | |
97.5 | |
0.8650g/mL | |
30°C | |
97.5% min. (GC) | |
C5H8O | |
(CH2CH)2CHOH | |
5g | |
1,4-pentadien-3-ol, ccris 8963, divinylcarbinol, divinyl carbinol, acmc-20ap1x, 1,4-pentadien-3-ol, contains hydroquinone as stabilizer | |
OC(C=C)C=C | |
84.12 | |
70204 | |
98% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
EINECSNumber : 213-080-9
RUO â Research Use Only