missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Oxathiane 4,4-dioxide, 99%

Manufacturer:  Alfa Aesar™ A1021006

 View more versions of this product

Catalog No. AAA1021006



Specifications

Specifications

1,4-Oxathiane 4,4-dioxide
131°C to 133°C
107-61-9
MFCD00011683
1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
C1CS(=O)(=O)CCO1
136.165
136.16
Odorless
5g
C4H8O3S
110609
WWRUZECKUVNAPB-UHFFFAOYSA-N
1,4-oxathiane 4,4-dioxide
7878
99%

Chemical Identifiers

107-61-9
136.165
WWRUZECKUVNAPB-UHFFFAOYSA-N
7878
C1CS(=O)(=O)CCO1
C4H8O3S
MFCD00011683
1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
1,4-oxathiane 4,4-dioxide
Safety and Handling

Safety and Handling

RTECSNumber : RP4375000

TSCA : TSCA

SDS Documents