missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,4-Oxathiane 4,4-dioxide, 99%

$45.18 - $437.96

Chemical Identifiers

CAS 107-61-9
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.165
MDL Number MFCD00011683
InChI Key WWRUZECKUVNAPB-UHFFFAOYSA-N
Synonym 1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
PubChem CID 7878
IUPAC Name 1,4-oxathiane 4,4-dioxide
SMILES C1CS(=O)(=O)CCO1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1021006 View Documents Alfa Aesar™
A1021006
5g Each for $45.18
 
AAA1021014 View Documents Alfa Aesar™
A1021014
25g Each for $128.00
 
AAA1021022 View Documents Alfa Aesar™
A1021022
100g Each for $437.96
 
Specifications

Chemical Identifiers

107-61-9
136.165
WWRUZECKUVNAPB-UHFFFAOYSA-N
7878
C1CS(=O)(=O)CCO1
C4H8O3S
MFCD00011683
1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
1,4-oxathiane 4,4-dioxide

Specifications

Odorless
5g
C4H8O3S
110609
WWRUZECKUVNAPB-UHFFFAOYSA-N
1,4-oxathiane 4,4-dioxide
7878
99%
131°C to 133°C
107-61-9
MFCD00011683
1,4-thioxane-1,1-dioxide, thioxane sulfone, p-thioxane sulfone, 1,4-oxathiane, 4,4-dioxide, oxathiane 4,4-dioxide, usaf do-38, 4,4-dioxo-1,4-oxathiane, 1,4-thioxane 1,1-dioxide, 1,4lambda6-oxathiane-4,4-dione, thioxanesulfone
C1CS(=O)(=O)CCO1
136.165
136.16
1,4-Oxathiane 4,4-dioxide
Safety and Handling

Safety and Handling

RTECSNumber : RP4375000

TSCA : TSCA