Learn More
1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2460022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(4-Nitrophenylazo)-2-naphthol | |
248°C to 252°C | |
MFCD00003908 | |
680469 | |
ACHQDMGGRZPFAW-VLGSPTGOSA-N | |
(1E)-1-[(4-nitrophenyl)hydrazinylidene]naphthalen-2-one | |
6076344 |
6410-10-2 | |
C16H11N3O3 | |
100 g | |
pigment red 1, federal red, para toner, label red, eljon para red, para red light, para red toner, para toner red, pigment red zh, lutetia red b | |
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)[N+](=O)[O-] | |
293.28 | |
293.28 |
Chemical Identifiers
6410-10-2 | |
293.28 | |
ACHQDMGGRZPFAW-VLGSPTGOSA-N |
C16H11N3O3 | |
MFCD00003908 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 229-093-8
RTECSNumber : QL4510000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only