Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Nitrophenyl)piperidine, 97%

$86.19 - $86.19

Chemical Identifiers

Chemical Name or Material 1-(4-Nitrophenyl)piperidine
CAS 6574-15-8
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD00023662
Synonym 1-Nitro-4-(1-piperidinyl)benzene; 1-Nitro-4-(1-piperidinyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2039906

  SDS
alfa aesar™
B2039906
5g Each for $86.19
Description & Specifications

Specifications

Formula Weight 206.24
Quantity 5g
Assay 97%
Molecular Formula C11H14N2O2
Melting Point 102° to 105°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-(4-Nitrophenyl)piperidine
MDL Number MFCD00023662
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Nitro-4-(1-piperidinyl)benzene; 1-Nitro-4-(1-piperidinyl)benzene
CAS 6574-15-8
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.