missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Nitrophenyl)azepane, 97%

$25.91 - $336.18

Chemical Identifiers

CAS 13663-23-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD00156364
InChI Key WXAAQKMTSQDMII-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl azepane, 1h-azepine, hexahydro-1-4-nitrophenyl, 1-4-nitro-phenyl-azepane, n-4'-nitrophenyl hexamethylenimine, hexahydro-1-4-nitrophenyl-1h-azepine, 4-nitrophenyl azaperhydroepine, 1-4-nitrophenyl-perhydroazepin, 1-4-nitrophenyl hexahydro-1h-azepine
PubChem CID 83639
IUPAC Name 1-(4-nitrophenyl)azepane
SMILES C1CCCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63887MD View Documents Alfa Aesar™
H63887MD
250mg Each for $25.91
 
AAH6388703 View Documents Alfa Aesar™
H6388703
1g Each for $76.65
 
AAH6388706 View Documents Alfa Aesar™
H6388706
5g Each for $336.18
 
Specifications

Chemical Identifiers

13663-23-5
220.272
WXAAQKMTSQDMII-UHFFFAOYSA-N
83639
C1CCCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
C12H16N2O2
MFCD00156364
1-4-nitrophenyl azepane, 1h-azepine, hexahydro-1-4-nitrophenyl, 1-4-nitro-phenyl-azepane, n-4'-nitrophenyl hexamethylenimine, hexahydro-1-4-nitrophenyl-1h-azepine, 4-nitrophenyl azaperhydroepine, 1-4-nitrophenyl-perhydroazepin, 1-4-nitrophenyl hexahydro-1h-azepine
1-(4-nitrophenyl)azepane

Specifications

250mg
C12H16N2O2
UN3259
WXAAQKMTSQDMII-UHFFFAOYSA-N
1-(4-nitrophenyl)azepane
83639
Solid
1-(4-Nitrophenyl)azepane
13663-23-5
MFCD00156364
1-4-nitrophenyl azepane, 1h-azepine, hexahydro-1-4-nitrophenyl, 1-4-nitro-phenyl-azepane, n-4'-nitrophenyl hexamethylenimine, hexahydro-1-4-nitrophenyl-1h-azepine, 4-nitrophenyl azaperhydroepine, 1-4-nitrophenyl-perhydroazepin, 1-4-nitrophenyl hexahydro-1h-azepine
C1CCCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
220.272
220.27
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : TSCA