Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%

$53.11 - $167.78

Chemical Identifiers

Chemical Name or Material 1-(4-Nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
CAS 142818-03-9
Molecular Formula C11H6F3N3O4
Formula Weight 301.19
MDL Number MFCD00173867
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2736903

 
alfa aesar™
H2736903
1g Each for $53.11

AAH2736906

 
alfa aesar™
H2736906
5g Each for $167.78
Description & Specifications

Specifications

Formula Weight 301.19
Chemical Name or Material 1-(4-Nitrophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Assay 97%
Molecular Formula C11H6F3N3O4
MDL Number MFCD00173867
CAS 142818-03-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 200° to 204°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.