Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Nitrophenyl)-3-(2-thienyl)-2-propen-1-one, 98%

$136.81 - $534.91

Chemical Identifiers

Chemical Name or Material 1-(4-Nitrophenyl)-3-(2-thienyl)-2-propen-1-one
CAS 6028-92-8
Molecular Formula C13H9NO3S
Formula Weight 259.29
MDL Number MFCD00022531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1337106

  SDS
alfa aesar™
A1337106
5g Each for $136.81

AAA1337114

  SDS
alfa aesar™
A1337114
25g Each for $534.91
Description & Specifications

Specifications

Formula Weight 259.29
Quantity 5g
MDL Number MFCD00022531
Melting Point 168° to 170°C
Chemical Name or Material 1-(4-Nitrophenyl)-3-(2-thienyl)-2-propen-1-one
CAS 6028-92-8
Molecular Formula C13H9NO3S
Assay 98%