missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Nitrophenyl)-2-piperidone, TCI America™

$79.45 - $270.42

Chemical Identifiers

CAS 38560-30-4
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD04037154
InChI Key VPCQXIGQMFWXII-UHFFFAOYSA-N
PubChem CID 22324463
IUPAC Name 1-(4-nitrophenyl)piperidin-2-one
SMILES C1CCN(C(=O)C1)C2=CC=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09751G View Documents TCI America
N09751G
1g Each for $79.45
 
N09755G View Documents TCI America
N09755G
5g Each for $270.42
 
Specifications

Chemical Identifiers

38560-30-4
220.228
VPCQXIGQMFWXII-UHFFFAOYSA-N
1-(4-nitrophenyl)piperidin-2-one
C11H12N2O3
MFCD04037154
22324463
C1CCN(C(=O)C1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Yellow
1g
C11H12N2O3
VPCQXIGQMFWXII-UHFFFAOYSA-N
1-(4-nitrophenyl)piperidin-2-one
22324463
Crystal-Powder at 20°C
1-(4-Nitrophenyl)-2-piperidone
99°C
38560-30-4
MFCD04037154
C1CCN(C(=O)C1)C2=CC=C(C=C2)[N+](=O)[O-]
220.228
220.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No