missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Nitrobenzyl)-1,2,4-triazole, TCI America™

$183.34 - $632.52

Chemical Identifiers

CAS 119192-09-5
Molecular Formula C9H8N4O2
Molecular Weight (g/mol) 204.189
MDL Number MFCD03840588
InChI Key NVRYCUYVBBCXHT-UHFFFAOYSA-N
PubChem CID 6482000
IUPAC Name 1-[(4-nitrophenyl)methyl]-1,2,4-triazole
SMILES C1=CC(=CC=C1CN2C=NC=N2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N100725G View Documents TCI America
N100725G
25g Each for $632.52
 
N10075G View Documents TCI America
N10075G
5g Each for $183.34
 
Specifications

Chemical Identifiers

119192-09-5
204.189
NVRYCUYVBBCXHT-UHFFFAOYSA-N
1-[(4-nitrophenyl)methyl]-1,2,4-triazole
C9H8N4O2
MFCD03840588
6482000
C1=CC(=CC=C1CN2C=NC=N2)[N+](=O)[O-]

Specifications

Undesignated
25g
C9H8N4O2
NVRYCUYVBBCXHT-UHFFFAOYSA-N
1-[(4-nitrophenyl)methyl]-1,2,4-triazole
6482000
Crystal-Powder at 20°C
1-(4-Nitrobenzyl)-1,2,4-triazole
102°C
119192-09-5
MFCD03840588
C1=CC(=CC=C1CN2C=NC=N2)[N+](=O)[O-]
204.189
204.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No