missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,4-Naphthalenebis(trifluoromethanesulfonate), TCI America™

$224.59 - $763.91

Chemical Identifiers

CAS 152873-78-4
Molecular Formula C12H6F6O6S2
Molecular Weight (g/mol) 424.284
InChI Key WUQONKZHJZYVAX-UHFFFAOYSA-N
Synonym 1,4-Naphthaleneditriflate
PubChem CID 10526326
IUPAC Name [4-(trifluoromethylsulfonyloxy)naphthalen-1-yl] trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C(=CC=C2OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09345G View Documents TCI America
N09345G
5g Each for $763.91
 
N09341G View Documents TCI America
N09341G
1g Each for $224.59
 
Specifications

Chemical Identifiers

152873-78-4
424.284
1,4-Naphthaleneditriflate
[4-(trifluoromethylsulfonyloxy)naphthalen-1-yl] trifluoromethanesulfonate
C12H6F6O6S2
WUQONKZHJZYVAX-UHFFFAOYSA-N
10526326
C1=CC=C2C(=C1)C(=CC=C2OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

White
5g
C12H6F6O6S2
WUQONKZHJZYVAX-UHFFFAOYSA-N
[4-(trifluoromethylsulfonyloxy)naphthalen-1-yl] trifluoromethanesulfonate
10526326
Crystal-Powder at 20°C
1,4-Naphthalenebis(trifluoromethanesulfonate)
64°C
152873-78-4
1,4-Naphthaleneditriflate
C1=CC=C2C(=C1)C(=CC=C2OS(=O)(=O)C(F)(F)F)OS(=O)(=O)C(F)(F)F
424.284
424.28
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator