Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Morpholinyl)-2-propanol, 98%

$84.82 - $293.82

Chemical Identifiers

Chemical Name or Material 1-(4-Morpholinyl)-2-propanol
CAS 2109-66-2
Molecular Formula C7H15NO2
Formula Weight 145.2
MDL Number MFCD00023378
Beilstein 1443
Synonym N-(2-Hydroxypropyl)morpholine; alpha-Methyl-4-morpholineethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1185909

  SDS
alfa aesar™
L1185909
10g Each for $84.82

AAL1185918

  SDS
alfa aesar™
L1185918
50g Each for $293.82
Description & Specifications

Specifications

Chemical Name or Material 1-(4-Morpholinyl)-2-propanol
CAS 2109-66-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 98%
Molecular Formula C7H15NO2
Formula Weight 145.2
MDL Number MFCD00023378
Beilstein 1443
Synonym N-(2-Hydroxypropyl)morpholine; alpha-Methyl-4-morpholineethanol
Boiling Point 218°C
Flash Point 92°C (197°F)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.