missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-[4-(methylsulfonyl)phenyl]ethan-1-one, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C9H10O3S
Linear Formula H3CC(O)C6H4SO2CH3
Formula Weight 198.24
Chemical Name or Material 1-[4-(Methylsulfonyl)phenyl]ethan-1-one
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
JFD01520EA SDS Fisher Scientific Company
JFD01520EA
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
H3CC(O)C6H4SO2CH3
1-[4-(Methylsulfonyl)phenyl]ethan-1-one
C9H10O3S
198.24
97%

Specifications

10g
97%
H3CC(O)C6H4SO2CH3
1-[4-(Methylsulfonyl)phenyl]ethan-1-one
10297-73-1
C9H10O3S
198.24
97%