Please login to your online account to display your discounted pricing

1-[4-(methylsulfonyl)phenyl]ethan-1-one, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  JFD01520EA

 View more versions of this product

Catalog No. JFD01520EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 198.24
Quantity 10g
Assay 97%
Linear Formula H3CC(O)C6H4SO2CH3
Molecular Formula C9H10O3S
Assay Percent Range 97%
Chemical Name or Material 1-[4-(Methylsulfonyl)phenyl]ethan-1-one
CAS 10297-73-1

Specifications

Chemical Name or Material 1-[4-(Methylsulfonyl)phenyl]ethan-1-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H10O3S
Linear Formula H3CC(O)C6H4SO2CH3
Formula Weight 198.24