Please login to your online account to display your discounted pricing

1-(4-Methylphenyl)piperazine dihydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(4-Methylphenyl)piperazine dihydrochloride
CAS 13078-14-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H16N2·2HCl
Formula Weight 249.18
MDL Number MFCD00035286
Synonym N-(p-Tolyl)piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC139180050

  SDS
thermo fisher scientific
AC139180050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 249.18
Quantity 5g
Color White
Melting Point 237° to 241°C
CAS Max % 100.0
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 13078-14-3
Chemical Name or Material 1-(4-Methylphenyl)piperazine dihydrochloride
Assay Percent Range 98%
MDL Number MFCD00035286
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym N-(p-Tolyl)piperazine dihydrochloride
Molecular Formula C11H16N2·2HCl
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.