Please login to your online account to display your discounted pricing

1-(4-Methylphenyl)piperazine, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC370830250

 View more versions of this product

Catalog No. AC370830250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Low Melting Solid
Formula Weight 176.26
Quantity 25g
Color White to yellow
Melting Point 29° to 34°C
CAS Max % 100.0
Chemical Name or Material 1-(4-Methylphenyl)piperazine
Molecular Formula C11H16N2
MDL Number MFCD00040737
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
CAS 39593-08-3
Synonym N-(p-Tolyl)piperazine
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

Specifications

Chemical Name or Material 1-(4-Methylphenyl)piperazine
CAS 39593-08-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD00040737
Synonym N-(p-Tolyl)piperazine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents