Learn More
1-(4-Methylphenyl)piperazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 370830050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-(4-Methylphenyl)piperazine | |
97.5 | |
Authentic | |
Glass bottle | |
MFCD00040737 | |
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
CC1=CC=C(C=C1)N2CCNCC2 | |
176.26 | |
176.26 |
39593-08-3 | |
100.0 | |
97.5% min. (GC) | |
C11H16N2 | |
5g | |
ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
1-(4-methylphenyl)piperazine | |
83113 | |
98% |
Chemical Identifiers
39593-08-3 | |
176.26 | |
ONEYFZXGNFNRJH-UHFFFAOYSA-N | |
83113 | |
CC1=CC=C(C=C1)N2CCNCC2 |
C11H16N2 | |
MFCD00040737 | |
1-4-methylphenyl piperazine, 1-p-tolyl piperazine, 1-p-tolyl-piperazine, 1-4-methylphenyl-piperazine, 1-4-methyl phenyl piperazine, 1-p-tolylpiperazine, acmc-1adtx, 4-methylphenyl piperazine, 4-4-methylphenyl piperazine, 1-4-methyl-phenyl-piperazine | |
1-(4-methylphenyl)piperazine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 254-534-6
RUO â Research Use Only