Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methylphenyl)homopiperazine monohydrochloride, 98%

$668.05 - $668.05

Chemical Identifiers

Chemical Name or Material 1-(4-Methylphenyl)homopiperazine monohydrochloride
CAS 1192191-39-1
Molecular Formula C12H18N2·HCl
Formula Weight 226.75
MDL Number MFCD16172201
Beilstein 19720425
Synonym 4-MPHP·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51750ME

  SDS
alfa aesar™
H51750ME
500mg Each for $668.05
Description & Specifications

Specifications

Formula Weight 226.75
Chemical Name or Material 1-(4-Methylphenyl)homopiperazine monohydrochloride
Assay 98%
Molecular Formula C12H18N2·HCl
MDL Number MFCD16172201
CAS 1192191-39-1
Beilstein 19720425
Synonym 4-MPHP·HCl
Melting Point 177° to 181°C
Quantity 500mg

Applications
1-(4-Methylphenyl)homopiperazine monohydrochloride is useful intermediate for synthesis.

Notes
Sold in collaboration with Tosoh Organic Chemical Co., Ltd.