Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methylphenyl)ethanol, 97%

$44.31 - $44.31

Chemical Identifiers

Chemical Name or Material 1-(4-Methylphenyl)ethanol
CAS 536-50-5
Molecular Formula C9H12O
Formula Weight 136.2
MDL Number MFCD00016860
Beilstein 1857562
Merck Index 14,3237
Synonym alpha,4-Dimethylbenzyl alcohol; 1-(p-Tolyl)ethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2083203

  SDS
alfa aesar™
B2083203
1g Each for $44.31
Description & Specifications

Specifications

Formula Weight 136.2
CAS 536-50-5
Chemical Name or Material 1-(4-Methylphenyl)ethanol
Flash Point 107°C (225°F)
Assay 97%
Merck Index 14,3237
Molecular Formula C9H12O
Quantity 1g
TSCA TSCA
RTECS Number DO4565000
MDL Number MFCD00016860
Synonym alpha,4-Dimethylbenzyl alcohol; 1-(p-Tolyl)ethanol
Beilstein 1857562
Boiling Point 218° to 220°C

Applications
1-(4-Methylphenyl)ethanol serves as a component of the essential oil from some plants in the ginger family. It is also used as a flavoring agent.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and strong reducing agents. Store in a cool place.