missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Methylphenoxy)-2-propanol 90.0+%, TCI America™

Manufacturer:  Tci America M106525G

 View more versions of this product

Catalog No. M106525G



Specifications

Specifications

1-(4-Methylphenoxy)-2-propanol
White-Yellow
4369-08-8
MFCD00191544
VTOVIJQMFYQNSX-UHFFFAOYNA-N
1-(4-methylphenoxy)propan-2-ol
12384823
Crystal-Lump at 20°C
130°C
25g
C10H14O2
1-(p-Tolyloxy)-2-propanol
CC1=CC=C(C=C1)OCC(C)O
166.22
166.22
≥90.0% (GC)

Chemical Identifiers

4369-08-8
166.22
VTOVIJQMFYQNSX-UHFFFAOYNA-N
12384823
CC1=CC=C(C=C1)OCC(C)O
C10H14O2
MFCD00191544
1-(p-Tolyloxy)-2-propanol
1-(4-methylphenoxy)propan-2-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions