missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methylbenzoyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H56037MD

 View more versions of this product

Catalog No. AAH56037MD



Description

Description

Specifications

Specifications

1-(4-Methylbenzoyl)pyrrolidine
59746-40-6
MFCD00996306
QXMOFZGGGOFRPH-UHFFFAOYSA-N
(4-methylphenyl)-pyrrolidin-1-ylmethanone
721203
97%
250mg
C12H15NO
1-4-methylbenzoyl pyrrolidine, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, 4-methylphenyl pyrrolidin-1-yl methanone, 4-methylphenyl pyrrolidinyl ketone, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-pyrrolidinecarbonyl toluene, aronis25719
CC1=CC=C(C=C1)C(=O)N2CCCC2
189.258
189.25

Chemical Identifiers

59746-40-6
189.258
QXMOFZGGGOFRPH-UHFFFAOYSA-N
721203
CC1=CC=C(C=C1)C(=O)N2CCCC2
C12H15NO
MFCD00996306
1-4-methylbenzoyl pyrrolidine, 1-4-methylphenyl carbonyl pyrrolidine, 4-methylphenyl 1-pyrrolidinyl methanone, 4-methylphenyl pyrrolidin-1-yl methanone, 4-methylphenyl pyrrolidinyl ketone, 4-methylphenyl-pyrrolidin-1-ylmethanone, 4-pyrrolidinecarbonyl toluene, aronis25719
(4-methylphenyl)-pyrrolidin-1-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents