missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methylbenzoyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H5603703

 View more versions of this product

Catalog No. AAH5603703



Specifications

Specifications

1-(4-Methylbenzoyl)pyrrolidine
59746-40-6
MFCD00996306
QXMOFZGGGOFRPH-UHFFFAOYSA-N
(4-methylphenyl)-pyrrolidin-1-ylmethanone
721203
97%
1g
C12H15NO
1-4-methylbenzoyl pyrrolidine
CC1=CC=C(C=C1)C(=O)N2CCCC2
189.258
189.25

Chemical Identifiers

59746-40-6
189.258
QXMOFZGGGOFRPH-UHFFFAOYSA-N
721203
CC1=CC=C(C=C1)C(=O)N2CCCC2
C12H15NO
MFCD00996306
1-4-methylbenzoyl pyrrolidine
(4-methylphenyl)-pyrrolidin-1-ylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents