missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methoxyphenylsulfonyl)pyrrolidine, 97%

Manufacturer:  Alfa Aesar™ H56035MD

 View more versions of this product

Catalog No. AAH56035MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-(4-Methoxyphenylsulfonyl)pyrrolidine
335215-12-8
MFCD00783727
LBMXZZFSJLBRGH-UHFFFAOYSA-N
1-(4-methoxyphenyl)sulfonylpyrrolidine
247207
97%
250mg
C11H15NO3S
1-4-methoxy-benzenesulfonyl-pyrrolidine
COC1=CC=C(C=C1)S(=O)(=O)N2CCCC2
241.305
241.31

Chemical Identifiers

335215-12-8
241.305
LBMXZZFSJLBRGH-UHFFFAOYSA-N
247207
COC1=CC=C(C=C1)S(=O)(=O)N2CCCC2
C11H15NO3S
MFCD00783727
1-4-methoxy-benzenesulfonyl-pyrrolidine
1-(4-methoxyphenyl)sulfonylpyrrolidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents