Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methoxyphenyl)piperazine dihydrochloride, 97%

$173.93 - $173.93

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)piperazine dihydrochloride
CAS 38869-47-5
Molecular Formula C11H16N2O·2HCl
Formula Weight 265.18
MDL Number MFCD00012767
Beilstein 4275784
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0200714

 
alfa aesar™
L0200714
25g Each for $173.93
Description & Specifications

Specifications

Formula Weight 265.18
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine dihydrochloride
Assay 97%
Molecular Formula C11H16N2O·2HCl
MDL Number MFCD00012767
CAS 38869-47-5
Melting Point ≈240°C
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 4275784
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.