missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(4-Methoxyphenyl)piperazine, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 255832500
Specifications
1-(4-Methoxyphenyl)piperazine | |
Glass bottle | |
250g | |
38212-30-5 | |
100.0 | |
C11H16N2O | |
1-4-anisyl piperazine, 1-4-methoxy-phenyl-piperazine, 1-4-methoxyphenyl piperazine, 1-4-methoxyphenyl-piperazine, 4-methoxyphenylpiperazine, chembl59517, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg | |
COC1=CC=C(C=C1)N2CCNCC2 | |
192.25 | |
192.25 | |
96% |
Authentic | |
Amber to Yellow | |
96% | |
95.0 | |
95% min. (GC) | |
MFCD00040736 | |
MRDGZSKYFPGAKP-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)piperazine | |
269722 | |
Crystalline Low Melting Solid |
Chemical Identifiers
38212-30-5 | |
192.25 | |
MRDGZSKYFPGAKP-UHFFFAOYSA-N | |
269722 | |
COC1=CC=C(C=C1)N2CCNCC2 |
C11H16N2O | |
MFCD00040736 | |
1-4-anisyl piperazine, 1-4-methoxy-phenyl-piperazine, 1-4-methoxyphenyl piperazine, 1-4-methoxyphenyl-piperazine, 4-methoxyphenylpiperazine, chembl59517, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg | |
1-(4-methoxyphenyl)piperazine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 253-829-7
TSCA : TSCA