missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)piperazine, 96%, ACROS Organics™

$403.37 - $403.37

Chemical Identifiers

CAS 38212-30-5
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C11H16N2O
Formula Weight 192.25
MDL Number MFCD00040736
Chemical Name or Material 1-(4-Methoxyphenyl)piperazine, 96%
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC255832500 SDS Thermo Fisher Scientific
255832500
Glass bottle 250g Each for $403.37
AC255830250 SDS Thermo Fisher Scientific
AC255830250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

38212-30-5
100.0
C11H16N2O
MFCD00040736
≥95%
95.0
96%
192.25
1-(4-Methoxyphenyl)piperazine, 96%

Specifications

38212-30-5
100.0
Authentic
96%
192.25
Light yellow to amber
1-(4-Methoxyphenyl)piperazine, 96%
95.0
Glass bottle
Crystalline Low Melting Solid
C11H16N2O
MFCD00040736
250g
≥95%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 253-829-7

TSCA : TSCA