Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methoxyphenyl)homopiperazine monohydrochloride, 98%

$500.03 - $500.03

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)homopiperazine monohydrochloride
CAS 934992-02-6
Molecular Formula C12H18N2O·HCl
Formula Weight 242.74g/mol
MDL Number MFCD11501072
Synonym 4-HPA·HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51703ME

 
alfa aesar™
H51703-ME
500mg Each for $500.03
Description & Specifications

Specifications

Quantity 500mg
Melting Point 175° to 179°C
Quantity 500mg
Chemical Name or Material 1-(4-Methoxyphenyl)homopiperazine monohydrochloride
CAS 934992-02-6
Assay 98%
Molecular Formula C12H18N2O·HCl
Formula Weight 242.74g/mol
MDL Number MFCD11501072
Synonym 4-HPA·HCl
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.