Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene, 99+%

$54.55 - $287.80

Chemical Identifiers

Chemical Name or Material 1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene
CAS 39969-28-3
Molecular Formula C20H22O
Formula Weight 278.39
MDL Number MFCD00187208
Synonym 1-(4-Methoxyphenyl)-2-(4-n-pentylphenyl)acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2788903

  SDS
alfa aesar™
H2788903
1g Each for $54.55

AAH2788909

  SDS
alfa aesar™
H2788909
10g Each for $287.80
Description & Specifications

Specifications

Formula Weight 278.39
Melting Point 47°C
Chemical Name or Material 1-[(4-Methoxyphenyl)ethynyl]-4-n-pentylbenzene
Molecular Formula C20H22O
MDL Number MFCD00187208
Assay ≥99%
Quantity 1g
Boiling Point 168° to 170°C (0.5mmHg)
Synonym 1-(4-Methoxyphenyl)-2-(4-n-pentylphenyl)acetylene
CAS 39969-28-3