Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methoxyphenyl)-1H-1,2,4-triazole, 99%

$96.88 - $322.45

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)-1H-1,2,4-triazole
CAS 68377-33-3
Molecular Formula C9H9N3O
Formula Weight 175.19
MDL Number MFCD12407079
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3096206

  SDS
alfa aesar™
H3096206
5g Each for $96.88

AAH3096214

  SDS
alfa aesar™
H3096214
25g Each for $322.45
Description & Specifications

Specifications

Formula Weight 175.19
Chemical Name or Material 1-(4-Methoxyphenyl)-1H-1,2,4-triazole
Assay 99%
Molecular Formula C9H9N3O
MDL Number MFCD12407079
CAS 68377-33-3
Melting Point 96° to 98°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.