Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C11H12O3
Formula Weight 192.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170710100

  SDS
fisher scientific acros
170710100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC170710500

  SDS
thermo fisher scientific
AC170710500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid
Color White to almost white
Melting Point 127° to 132°C
Quantity 10g
CAS 16728-01-1
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C11H12O3
Formula Weight 192.21