Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170740250

 View more versions of this product

Catalog No. AC170740250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 220.27
Quantity 25g
Color Beige
EINECS Number 256-063-1
TSCA TSCA
Melting Point 151° to 156°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%
Assay Percent Range 95%
Molecular Formula C13H16O3
Linear Formula CH3OC6H4C5H8CO2H
MDL Number MFCD00001374
CAS 43050-28-8

Specifications

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid
Name Note 95%
CAS 43050-28-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16O3
Linear Formula CH3OC6H4C5H8CO2H
Formula Weight 220.27
MDL Number MFCD00001374