Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid, 95%, ACROS Organics™

$153.08 - $153.08

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid
Name Note 95%
CAS 43050-28-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C13H16O3
Linear Formula CH3OC6H4C5H8CO2H
Formula Weight 220.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170740250

  SDS
thermo fisher scientific
170740250
Glass bottle 25g Each for $153.08
Description & Specifications

Specifications

Assay ≥94%
Physical Form Powder
Formula Weight 220.27
Quantity 25g
Color Beige
EINECS Number 256-063-1
TSCA TSCA
Melting Point 151° to 156°C
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclopentanecarboxylic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 95%
Assay Percent Range 95%
Molecular Formula C13H16O3
Linear Formula CH3OC6H4C5H8CO2H
MDL Number MFCD00001374
CAS 43050-28-8