Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile, 95%, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  170670050

 View more versions of this product

Catalog No. AC170670050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 215.29
Physical Form Low Melting Solid
Quantity 5g
Color Colorless to yellow-brown
TSCA TSCA
EINECS Number 252-938-7
Density 1.0778g/mL
CAS Max % 100.0
CAS Min % 94.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
MDL Number MFCD00021282
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Molecular Formula C14H17NO
Assay Percent Range 95%
CAS 36263-51-1

Specifications

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Name Note 95%
CAS 36263-51-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H17NO
Formula Weight 215.29
MDL Number MFCD00021282
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents