Please login to your online account to display your discounted pricing

1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Name Note 95%
CAS 36263-51-1
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C14H17NO
Formula Weight 215.29
MDL Number MFCD00021282
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC170670050

  SDS
thermo fisher scientific
170670050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Low Melting Solid
Formula Weight 215.29
Quantity 5g
Color Colorless to yellow-brown
EINECS Number 252-938-7
TSCA TSCA
Density 1.0778g/mL
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Infrared Spectrum Authentic
Name Note 95%
Assay Percent Range 95%
Molecular Formula C14H17NO
MDL Number MFCD00021282
CAS 36263-51-1
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.