Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile, 94%

Chemical Identifiers

Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
CAS 36263-51-1
Molecular Formula C14H17NO
Formula Weight 215.3
MDL Number MFCD00021282
UN Number UN3439
Beilstein 2941304
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0610706

  SDS
alfa aesar™
L0610706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0610714

  SDS
alfa aesar™
L0610714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 215.3
Chemical Name or Material 1-(4-Methoxyphenyl)-1-cyclohexanecarbonitrile
Assay 94%
Molecular Formula C14H17NO
MDL Number MFCD00021282
CAS 36263-51-1
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3439
Beilstein 2941304
Melting Point 43° to 60°C
Quantity 5g
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.