Alfa Aesar™ 1-(4-Iodophenyl)pyrrole, 97%
Manufacturer:  Alfa Aesar™ A1530514
 View more versions of this product
Catalog No. AAA1530514
Specifications
Specifications
1-(4-Iodophenyl)pyrrole
25g
C10H8IN
7915717
FMURNAZHVQDQQN-UHFFFAOYSA-N
1-(4-iodophenyl)pyrrole
272430
97%
128°C to 133°C
92636-36-7
MFCD00052399
1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
C1=CN(C=C1)C2=CC=C(C=C2)I
269.085
269.09
Chemical Identifiers
92636-36-7
269.085
FMURNAZHVQDQQN-UHFFFAOYSA-N
272430
C1=CN(C=C1)C2=CC=C(C=C2)I
C10H8IN
MFCD00052399
1-4-iodophenyl pyrrole, 1-4-iodophenyl-1h-pyrrole, acmc-20apaj, 4-iodophenyl pyrrole, maybridge1_002926, 1-4-iodo-phenyl-1h-pyrrole, 1h-pyrrole,1-4-iodophenyl, 1-4-iodophenyl-1h-pyrrole #
1-(4-iodophenyl)pyrrole
Documents